6-Methyl-2-thiouracil 98

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Methiocil, 2-Mercapto-6-methylpyrimidin-4(1H)-one, Methylthiouracil, MZU, Basethyrin, 4-Hydroxy-2-mercapto 6-methylpyrimidine, Thiothymin

A17982.18 A17982.30 A17982.A1
A17982.18EA 38.9 CHF
A17982.18 A17982.30 A17982.A1
6-Methyl-2-thiouracil 98
6-Methyl-2-thiouracil

Purity: 98% CAS No.: 56-04-2 Molecular Formula: C5H6N2OS Formula Weight: 142.18 Melting Point: ca 329° dec. Beilstein Reference: 115648 Merck Reference: 14,6128 MDL No.: MFCD00006040

Formel: C₅H₆N₂OS
MW: 142.18 g/mol
Siedepunkt: 351
Melting Pt: ∼ 329 °C (Zers.)
Dichte: 1.519 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006040
CAS-Nummer: 56-04-2
EINECS: 200-252-3
Merck Index: 14,6128

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR