Phenylsilan ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Silylbenzene

A17901.06 A17901.14 A17901.22
A17901.06EA 138 CHF
A17901.06 A17901.14 A17901.22
Phenylsilan ≥97%
Phenylsilane

Purity: 97% CAS No.: 694-53-1 Molecular Formula: C6H8Si Formula Weight: 108.22 Flash Point: 7°(45°F) Refractive Index: 1.5105 Boiling Point: 120° UN No.: 3399 MDL No.: MFCD00013478

Formel: C₆H₈Si
MW: 108.22 g/mol
Siedepunkt: 120 °C
Dichte: 0.88
Flash-Pt: 7 °C (45 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013478
CAS-Nummer: 694-53-1
EINECS: 211-772-5
UN: 3399
ADR: 4.3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR