N,N-Diethyl-1,4-phenylendiammoniumsulfat 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, 4-Amino-N,N-diethylaniline sulphate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate

A17832.0B A17832.18 A17832.30
A17832.0BEA 574 CHF
A17832.0B A17832.18 A17832.30
N,N-Diethyl-1,4-phenylendiammoniumsulfat 97%
N,N-Diethyl-1,4-phenylendiammoniumsulfat

Purity: 97% CAS No.: 6283-63-2 Molecular Formula: C10H16N2.H2SO4 Formula Weight: 262.33 Melting Point: 183-188° Beilstein Reference: 4219768

Formel: C₁₀H₁₈N₂O₄S
Siedepunkt: 274…275 °C (1013 hPa)
Melting Pt: 183…188 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00012993
CAS-Nummer: 6283-63-2
EINECS: 228-500-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR