Amberlite® IRA-410 (Cl) Ionenaustauscher

Lieferant: Thermo Fisher Scientific

Synonyms: Amberlite® IRA-410 Cl type

A17734.0E A17734.22 A17734.36
A17734.0EEA 557 CHF
A17734.0E A17734.22 A17734.36
Amberlite® IRA-410 (Cl) Ionenaustauscher
Amberlite® IRA-410 (Cl)

CAS No.: 9002-26-0 Odour: Amine-like MDL No.: MFCD00132710

Lager Temperatur: Raumtemperatur
MDL Number: MFCD00132710
CAS-Nummer: 9002-26-0
Merck Index: 14,382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR