(+)-Cinchonin ≥98% cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol

A17523.14 A17523.22
A17523.14EA 87.9 CHF
A17523.14 A17523.22
(+)-Cinchonin ≥98% cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine
(+)-Cinchonin

Purity: 98+% CAS No.: 118-10-5 Molecular Formula: C19H22N2O Formula Weight: 294.39 Melting Point: 253-258° Beilstein Reference: 89689 Merck Reference: 14,2287 Optical Rotation: +225° (c=0.5 in ethanol) MDL No.: MFCD00064372

Formel: C₁₉H₂₂N₂O
MW: 294.4 g/mol
Melting Pt: 253…258 °C
Dichte: 1.2 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00064372
CAS-Nummer: 118-10-5
EINECS: 204-234-6
Merck Index: 13,02308

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR