tri-Ammoniumcitrat ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid triammonium salt, Ammonium citrate tribasic, Citric acid triammonium salt, Triammonium 2-hydroxypropane-1,2,3-tricarboxylate

A16973.22 A16973.36
A16973.22EA 127 CHF
A16973.22 A16973.36
tri-Ammoniumcitrat ≥97%
tri-Ammoniumcitrat

Purity: 97+% CAS No.: 3458-72-8 Molecular Formula: C6H17N3O7 Formula Weight: 243.22 Physical Form: Crystalline powder Appearance: White Melting Point: ca 180° dec. Solubility: Soluble in water MDL No.: MFCD00036406

Formel: C₆H₁₇N₃O₇
Melting Pt: ∼ 180 °C (dec.)
Dichte: 1 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00036406
CAS-Nummer: 3458-72-8
EINECS: 222-394-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR