Echtgrün FCF

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Light green, Sodium 2-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)cyclohexa-2,5-dien-1-ylidene)methyl)-5-hydroxybenzenesulphonate, CI 42053, Dihydrogen (ethyl)[4-[4-[ethyl(3-sulphonatobenzyl)amino](4-hydroxy-2-sulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene](3-sulphonatobenzyl)ammonium, disodium salt

A16520.06 A16520.22
A16520.06EA 62.6 CHF
A16520.06 A16520.22
Echtgrün FCF
Echtgrün FCF

CAS No.: 2353-45-9 Molecular Formula: C37H34N2Na2O10S3 Formula Weight: 808.86 Melting Point: ca 290° dec. Beilstein Reference: 5718212 Merck Reference: 14,3941 MDL No.: MFCD00013053

Formel: C₃₇H₃₄N₂Na₂O₁₀S₃
MW: 808.86 g/mol
Melting Pt: ∼ 290 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013053
CAS-Nummer: 2353-45-9
EINECS: 219-091-5
Merck Index: 13,03970

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR