Kaliumacetat 99

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Acetic acid potassium salt

A16321.0C A16321.0E A16321.36
A16321.0CEA 643 CHF
A16321.0C A16321.0E A16321.36
Kaliumacetat 99
Kaliumacetat

Purity: 99% CAS No.: 127-08-2 Molecular Formula: CH3CO2K Formula Weight: 98.15 Melting Point: 292° Beilstein Reference: 3595449 Merck Reference: 14,7605 MDL No.: MFCD00012458

Formel: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 292 °C
Dichte: 1.57
Flash-Pt: >250 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00012458
CAS-Nummer: 127-08-2
EINECS: 204-822-2
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR