Kupfer(II)acetat Monohydrat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Copper diacetate monohydrate, Cupric acetate monohydrate, Acetic acid copper (II) salt monohydrate, Copper(II) acetate hydrate

A16203.30 A16203.0B A16203.0I
A16203.30EA 74.2 CHF
A16203.30 A16203.0B A16203.0I
Kupfer(II)acetat Monohydrat ≥98%
Kupfer(II)acetat Monohydrat

Purity: 98+% CAS No.: 6046-93-1 Molecular Formula: C4H6CuO4.H2O Formula Weight: 199.65 (181.64anhy) Physical Form: Powder Density: 1.882 Appearance: Blue-green Odour: Odourless pH: 5.2-5.5 Melting Point: 115° Merck Reference: 14,2624 UN No.: 3077 MDL No.: MFCD00149570

Formel: (H₃CCOO)₂Cu·H₂O
MW: 199.65 g/mol
Melting Pt: 115
Dichte: 1.882
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00149570
CAS-Nummer: 6046-93-1
EINECS: 205-553-3
UN: 3077
ADR: 9 III
Merck Index: 13,02651

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR