Thymin 97%

Lieferant: Thermo Fisher Scientific

Synonyms: 5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil

A15879.14 A15879.22 A15879.36
A15879.14EA 108 CHF
A15879.14 A15879.22 A15879.36
Thymin 97%
Thymin

Purity: 97% CAS No.: 65-71-4 Molecular Formula: C5H6N2O2 Formula Weight: 126.12 Melting Point: 316-317° Beilstein Reference: 607626 Merck Reference: 14,9398 MDL No.: MFCD00006026

Formel: C₅H₆N₂O₂
MW: 126.11 g/mol
Siedepunkt: 389
Melting Pt: 316…317 °C
Dichte: 1.454 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006026
CAS-Nummer: 65-71-4
EINECS: 200-616-1
Merck Index: 12,09539

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR