Methylformiat ≥97% may cont. up to ca 3% methanol

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Formic acid methyl ester

A15850.0D A15850.0F A15850.AP
A15850.0DEA 349 CHF
A15850.0D A15850.0F A15850.AP
Methylformiat ≥97% may cont. up to ca 3% methanol
Methylformiat

Purity: 97% CAS No.: 107-31-3 Molecular Formula: C2H4O2 Formula Weight: 60.05 Storage Temperature: 2°C to 8°C Flash Point: -26°(-14°F) Physical Form: Clear liquid Density: 0.972 Refractive Index: 1.3440 Appearance: Colourless Melting Point: -100° Boiling Point: 32-33° Beilstein Reference: 1734623 Merck Reference: 14,6077 UN No.: 1243 MDL No.: MFCD00003291

Formel: C₂H₄O₂
Siedepunkt: 32…33 °C
Melting Pt: –100 °C
Dichte: 0.972
Flash-Pt: –26 °C (–14 °F)
Lager Temperatur: Kühlschrank
MDL Number: MFCD00003291
CAS-Nummer: 107-31-3
EINECS: 203-481-7
UN: 1243
ADR: 3,I
Merck Index: 14,6077

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR