Ethyloleat 70%, tech.

Lieferant: Thermo Fisher Scientific

Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester

A15601.0B A15601.0I A15601.30
A15601.0BEA 185 CHF
A15601.0B A15601.0I A15601.30
Ethyloleat 70%, tech.
Ethyloleat

Purity: 70% CAS No.: 111-62-6 Molecular Formula: C20H38O2 Formula Weight: 310.53 Flash Point: 110°(230°F) Refractive Index: 1.451 Melting Point: -32° Boiling Point: 205-208° Beilstein Reference: 1727318 MDL No.: MFCD00009579

Formel: C₂₀H₃₈O₂
MW: 310.52 g/mol
Siedepunkt: 205…208 °C
Melting Pt: –32 °C
Dichte: 0.87
Flash-Pt: >110 °C (230 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009579
CAS-Nummer: 111-62-6
EINECS: 203-889-5
Merck Index: 12,06965

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR