3-Chlor-p-toluidin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 3-Chloro-p-methylaniline, 2-Chloro-4-aminotoluene, 4-Amino-2-chlorotoluene, 3-Chloro-4-methylaniline

A15439.0B A15439.30
A15439.0BEA 420 CHF
A15439.0B A15439.30
3-Chlor-p-toluidin ≥98%
3-Chlor-p-toluidin

Purity: 98% CAS No.: 95-74-9 Molecular Formula: C7H8ClN Formula Weight: 141.60 Flash Point: 130°(266°F) Refractive Index: 1.5840 Melting Point: 22-24° Boiling Point: 236-238° Beilstein Reference: 636511 UN No.: 2239 MDL No.: MFCD00007773

Formel: C₇H₈ClN
Siedepunkt: 236…238 °C
Melting Pt: 22…24 °C
Dichte: 1.180
Flash-Pt: 130 °C (266 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007773
CAS-Nummer: 95-74-9
EINECS: 202-446-3
UN: 2239
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR