Aluminon

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt

A15401.09 A15401.18 A15401.30
A15401.09EA 56.3 CHF
A15401.09 A15401.18 A15401.30
Aluminon
Aluminon

CAS No.: 569-58-4 Molecular Formula: C22H23N3O9 Formula Weight: 473.44 Odour: Odourless pH: 5.3 Melting Point: 220-225° dec. MDL No.: MFCD00040925

Formel: C₂₂H₂₃N₃O₉
Melting Pt: 220…225 °C (dec.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00040925
CAS-Nummer: 569-58-4
EINECS: 209-319-1
Merck Index: 14,322

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR