Solvent Blue 38

Lieferant: Thermo Fisher Scientific

Synonyms: Direct Blue 86, C.I. 74180, Luxol® Fast Blue MBSN

A15395.14 A15395.22
A15395.14EA 77 CHF
A15395.14 A15395.22
Solvent Blue 38
Solvent Blue 38

Solvent Blue 38 is commonly utilized in microscopy to detect demyelination in the central nervous system. It is used as a myelin-sheath stain.


CAS No.: 1328-51-4 Odour: Odourless Melting Point: ca 235° dec. MDL No.: MFCD00071424

Formel: C32H12CuN8Na2O6S2
MW: 778.15 g/mol
Melting Pt: ∼ 235 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00071424
CAS-Nummer: 1328-51-4
EINECS: 215-523-1

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR