4-Fluorbenzaldehyd ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A15383.14 A15383.22 A15383.36
A15383.14EA 62 CHF
A15383.14 A15383.22 A15383.36
4-Fluorbenzaldehyd ≥98%
4-Fluorbenzaldehyd

Purity: 98% CAS No.: 459-57-4 Molecular Formula: C7H5FO Formula Weight: 124.11 Flash Point: 56°(133°F) Refractive Index: 1.5210 Melting Point: -10° Boiling Point: 180-181° Beilstein Reference: 385857 UN No.: 1989 MDL No.: MFCD00003378

Formel: C₇H₅FO
MW: 124.11 g/mol
Siedepunkt: 180…181 °C
Melting Pt: –10 °C
Dichte: 1.175
Flash-Pt: 56 °C (133 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003378
CAS-Nummer: 459-57-4
EINECS: 207-293-6
UN: 1989
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR