(4-Bromphenyl)essigsäure ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 4-Bromophenylacetic acid, 2-(4-bromophenyl)acetic acid

A15325.14 A15325.22
A15325.14EA 129 CHF
A15325.14 A15325.22
(4-Bromphenyl)essigsäure ≥99%
(4-Bromphenyl)essigsäure

Purity: 99% CAS No.: 1878-68-8 Molecular Formula: C8H7BrO2 Formula Weight: 215.05 Melting Point: 115-119° Beilstein Reference: 972146

Formel: (C₆H₄Br)CH₂COOH
MW: 215.05 g/mol
Siedepunkt: 326
Melting Pt: 115…119 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004342
CAS-Nummer: 1878-68-8
EINECS: 217-523-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR