1,3-Propandithiol 97

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 1,3-Dimercaptopropane

A15261.06 A15261.14 A15261.22
A15261.06EA 50.4 CHF
A15261.06 A15261.14 A15261.22
1,3-Propandithiol 97
1,3-Propandithiol

Purity: 97% CAS No.: 109-80-8 Molecular Formula: HS(CH2)3SH Formula Weight: 108.23 Flash Point: 63°(145°F) Refractive Index: 1.5400 Odour: Stench Melting Point: -79° Boiling Point: 168-170° Solubility: Slightly soluble in water. Miscible with alcohol, ether, chloroform, ether Beilstein Reference: 1071197 Merck Reference: 14,7801 UN No.: 3334 MDL No.: MFCD00004904

Formel: C₃H₈S₂
MW: 108.23 g/mol
Siedepunkt: 168…170 °C
Melting Pt: –79 °C
Dichte: 1.077
Flash-Pt: 63 °C (145 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004904
CAS-Nummer: 109-80-8
EINECS: 203-706-9
UN: 3334
ADR: 9 III
Merck Index: 12,07984

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR