Cholesteryl-acetat 97

Lieferant: Thermo Fisher Scientific

Synonyms: (3S,8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl acetate, Cholesteryl Acetate, Acetyl Cholesterol, Acetic Acid Cholesterol Ester

A15052.14 A15052.22 A15052.36
A15052.14EA 186 CHF
A15052.14 A15052.22 A15052.36
Cholesteryl-acetat 97
Cholesterol acetate

Purity: 97+% CAS No.: 604-35-3 Molecular Formula: C29H48O2 Formula Weight: 428.70 Melting Point: 112-114° Beilstein Reference: 2064235 Optical Rotation: -43° (c=5 in chloroform) MDL No.: MFCD00003636

Formel: C₂₉H₄₈O₂
MW: 428.70 g/mol
Melting Pt: 112…114 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003636
CAS-Nummer: 604-35-3
EINECS: 210-066-4

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR