Adenin 99

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Purin-6-amine, 7H-Purin-6-amine, 6-Aminopurine

A14906.14 A14906.18 A14906.30 A14906.36 A14906.A7
A14906.14EA 109 CHF
A14906.14 A14906.18 A14906.30 A14906.36 A14906.A7
Adenin 99
Adenin

Purity: 99% CAS No.: 73-24-5 Molecular Formula: C5H5N5 Formula Weight: 135.13 Odour: Odourless Melting Point: 360° dec. Beilstein Reference: 5777 Merck Reference: 14,152 UN No.: 2811 MDL No.: MFCD00041790

Formel: C₅H₅N₅
MW: 135.13 g/mol
Melting Pt: >360 °C (Zers.)
Dichte: 0.99741 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00041790
CAS-Nummer: 73-24-5
EINECS: 200-796-1
UN: 2811
ADR: 6.1 III
Merck Index: 13,00152

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR