2-Methylcyclohexanon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A14741.0F A14741.AE A14741.AP
A14741.0FEA 1550 CHF
A14741.0F A14741.AE A14741.AP
2-Methylcyclohexanon ≥98%
2-Methylcyclohexanone

Purity: 98% CAS No.: 583-60-8 Molecular Formula: C7H12O Formula Weight: 112.17 Flash Point: 46°(114°F) Physical Form: Clear liquid Density: 0.925 Refractive Index: 1.4480 Appearance: Colourless Melting Point: -14° Boiling Point: 162-163° Beilstein Reference: 506751 UN No.: 2297 MDL No.: MFCD00001635

Formel: C₇H₁₂O
Siedepunkt: 162…163 °C
Melting Pt: –14 °C
Dichte: 0.925
Flash-Pt: 46 °C (114 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001635
CAS-Nummer: 583-60-8
EINECS: 209-513-6
UN: 2297
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR