2-Oxazolidinon 99

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 2-Oxazolidone, Oxazolidin-2-one

A14543.14 A14543.22 A14543.36 AB118367.0025 AB118367.0100
A14543.14EA 87.7 CHF
A14543.14 A14543.22 A14543.36 ABCRAB118367.0025 ABCRAB118367.0100
2-Oxazolidinon 99
2-Oxazolidinon

Purity: 99% CAS No.: 497-25-6 Molecular Formula: C3H5NO2 Formula Weight: 87.08 Melting Point: 84-89° Boiling Point: 220°/48mm MDL No.: MFCD00005268

Formel: C₃H₅NO₂
MW: 87.08 g/mol
Siedepunkt: 220 °C (48 mmHg)
Melting Pt: 84…89 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005268
CAS-Nummer: 497-25-6
EINECS: 207-840-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR