Ethoxymethylenmalononitril ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

A14412.14 A14412.22 A14412.36
A14412.14EA 72.7 CHF
A14412.14 A14412.22 A14412.36
Ethoxymethylenmalononitril ≥97%
Ethoxymethylenmalononitril

Purity: 97+% CAS No.: 123-06-8 Molecular Formula: C6H6N2O Formula Weight: 122.13 Flash Point: 155°(311°F) Melting Point: 62-68° Boiling Point: 160°/12mm Beilstein Reference: 1634241 UN No.: 3439 MDL No.: MFCD00001854

Formel: C₆H₆N₂O
Siedepunkt: 160 °C (12 mmHg)
Melting Pt: 62…68 °C
Flash-Pt: 155 °C (311 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001854
CAS-Nummer: 123-06-8
EINECS: 204-597-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR