Ethylchlorfluoracetat ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Ethylchlorofluoroacetate

A14196.14
A14196.14EA 279 CHF
A14196.14
Ethylchlorfluoracetat ≥98%
Ethylchlorfluoracetat

Purity: 98% CAS No.: 401-56-9 Molecular Formula: ClFCHCO2CH2CH3 Formula Weight: 140.54 Flash Point: 44°(111°F) Refractive Index: 1.3960 Odour: Pungent Boiling Point: 128-130° Beilstein Reference: 1748679 UN No.: 2920 MDL No.: MFCD00039333

Formel: C₄H₆ClFO₂
MW: 140.54 g/mol
Siedepunkt: 128…130 °C
Dichte: 1.225
Flash-Pt: 44 °C (111 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00039333
CAS-Nummer: 401-56-9
EINECS: 206-930-5
UN: 2920
ADR: 8 II

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR