Trimethylorthoformiat ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A13760.0F A13760.AE A13760.AP
A13760.0FEA 412 CHF
A13760.0F A13760.AE A13760.AP
Trimethylorthoformiat ≥99%
Trimethylorthoformiat

Purity: 99% CAS No.: 149-73-5 Molecular Formula: HC(OCH3)3 Formula Weight: 106.12 Flash Point: 15°(59°F) Refractive Index: 1.3790 Melting Point: -53° Boiling Point: 101-102° Beilstein Reference: 969215 Merck Reference: 14,6884 UN No.: 3272 MDL No.: MFCD00008483

Formel: C₄H₁₀O₃
Siedepunkt: 101…102 °C
Melting Pt: –53 °C
Dichte: 0.968
Flash-Pt: 15 °C (59 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00008483
CAS-Nummer: 149-73-5
EINECS: 205-745-7
UN: 3272
ADR: 3,II
Merck Index: 14,6884

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR