p-Toluidin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 4-Aminotoluene, 4-Methylaniline, 4-Toluidine

A13698.0B A13698.0I A13698.30
A13698.0BEA 156 CHF
A13698.0B A13698.0I A13698.30
p-Toluidin ≥99%
p-Toluidin

Purity: 99+% CAS No.: 106-49-0 Molecular Formula: C7H9N Formula Weight: 107.16 Flash Point: 88°(190°F) Melting Point: 42-46° Boiling Point: 199-202° Beilstein Reference: 471281 Merck Reference: 14,9536 UN No.: 3451 MDL No.: MFCD00007906

Formel: C₇H₉N
Siedepunkt: 199…202 °C
Melting Pt: 42…46 °C
Dichte: 1.05
Flash-Pt: 88 °C (190 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007906
CAS-Nummer: 106-49-0
EINECS: 203-403-1
UN: 3451
ADR: 6.1,II
Merck Index: 14,9536

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR