myo-Inosit 98

Lieferant: Thermo Fisher Scientific

Synonyms: i-Inositol, D-Myo-Inositol, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol, Inositol

A13586.0E A13586.22 A13586.36
A13586.0EEA 1010 CHF
A13586.0E A13586.22 A13586.36
myo-Inosit 98
myo-Inosit

Purity: 98+% CAS No.: 87-89-8 Molecular Formula: C6H12O6 Formula Weight: 180.16 Physical Form: Cryst powder Density: 1.75 Appearance: White Odour: Odourless Melting Point: 224-227° Beilstein Reference: 1907329 Merck Reference: 14,4978 MDL No.: MFCD00077932


A lipotropic agent

Formel: C₆H₁₂O₆
MW: 180.16 g/mol
Siedepunkt: 291
Melting Pt: 224…227 °C
Dichte: 1.75
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00077932
CAS-Nummer: 87-89-8
EINECS: 201-781-2
Merck Index: 13,05001

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR