5-Fluoruracil 99

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: Fluorouracil, 5-Fluoropyrimidine-2,4(1H,3H)-dione, 5-FU

A13456.06 A13456.14 A13456.22
A13456.06EA 96.1 CHF
A13456.06 A13456.14 A13456.22
5-Fluoruracil 99
5-Fluoruracil

Purity: 99% CAS No.: 51-21-8 Molecular Formula: C4H3FN2O2 Formula Weight: 130.08 Melting Point: ca 282° dec. Beilstein Reference: 127172 Merck Reference: 14,4181 UN No.: 2811 MDL No.: MFCD00006018

Formel: C₄H₃FN₂O₂
MW: 130.08 g/mol
Siedepunkt: 347
Melting Pt: ∼ 282 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006018
CAS-Nummer: 51-21-8
EINECS: 200-085-6
UN: 2811
ADR: 6.1 III
Merck Index: 13,04208

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR