Aluminiumtri-sec-butanolat 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Aluminium Tri-sec-butylate, Aluminum s-butoxide, Aluminium tri-sec-butoxide

A13044.36 A13044.0E A13044.22
A13044.36EA 193 CHF
A13044.36 A13044.0E A13044.22
Aluminiumtri-sec-butanolat 97%
Aluminiumtri-sec-butanolat

Purity: 97% CAS No.: 2269-22-9 Molecular Formula: Al[OCH(CH3)CH2CH3]3 Formula Weight: 246.32 Flash Point: 27°(82°F) Refractive Index: 1.4390 Odour: Alcohol-like Boiling Point: 200-206°/30mm Beilstein Reference: 3910908 UN No.: 1992 MDL No.: MFCD00009327

Formel: C₁₂H₂₇AlO₃
MW: 246.33 g/mol
Siedepunkt: 200…206 °C (30 mmHg)
Dichte: 0.967
Flash-Pt: 27 °C (82 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00009327
CAS-Nummer: 2269-22-9
EINECS: 218-871-2
UN: 1992
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR