Trifluoressigsäure 99

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: TFA

A12198.22 A12198.36 A12198.0E A12198.A9 A12198.C4
A12198.22EA 92.5 CHF
A12198.22 A12198.36 A12198.0E A12198.A9 A12198.C4
Trifluoressigsäure 99
Trifluoressigsäure

Purity: 99% CAS No.: 76-05-1 Molecular Formula: CF3CO2H Formula Weight: 114.02 Physical Form: Liquid Density: 1.480 Refractive Index: 1.2840 Sensitivity: Hygroscopic Odour: Pungent Melting Point: -15° Boiling Point: 72-73° Solubility: Miscible with ether, acetone, ethanol, benzene, hexane, CCl4 Beilstein Reference: 742035 Merck Reference: 14,9681 UN No.: 2699 MDL No.: MFCD00004169


Solvent, catalyst, strong non-oxidizing acid

Formel: C₂HF₃O₂
MW: 114.02 g/mol
Siedepunkt: 72…73 °C
Melting Pt: –15 °C
Dichte: 1.480
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00004169
CAS-Nummer: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8 I
Merck Index: 14,9681
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR