2',5'-Dihydroxyacetophenon ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 1-(2,5-Dihydroxyphenyl)ethan-1-one, 1-(2,5-Dihydroxyphenyl)ethanone, 2,5-Dihydroxyacetophenone

A12185.06 A12185.14 A12185.22
A12185.06EA 69.2 CHF
A12185.06 A12185.14 A12185.22
2',5'-Dihydroxyacetophenon ≥98%
2',5'-Dihydroxyacetophenon

Purity: 98+% CAS No.: 490-78-8 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 204-208° Beilstein Reference: 637903

Formel: C₈H₈O₃
MW: 152.15 g/mol
Siedepunkt: 296
Melting Pt: 204…208 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00002343
CAS-Nummer: 490-78-8
EINECS: 207-716-4

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR