α-Brom-2-chlortoluol ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 1-(Bromomethyl)-2-chlorobenzene, 2-Chlorobenzyl bromide

A12129.06 A12129.14 A12129.22
A12129.06EA 64.3 CHF
A12129.06 A12129.14 A12129.22
α-Brom-2-chlortoluol ≥97%
α-Brom-2-chlortoluol

Purity: 97% CAS No.: 611-17-6 Molecular Formula: C7H6BrCl Formula Weight: 205.49 Flash Point: 108°(226°F) Refractive Index: 1.5920 Odour: Pungent Boiling Point: 103-104°/10mm Beilstein Reference: 386264 UN No.: 3265 MDL No.: MFCD00000566

Formel: C₇H₆BrCl
MW: 205.48 g/mol
Siedepunkt: 103…104 °C (10 mmHg)
Dichte: 1.583
Flash-Pt: 108 °C (226 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000566
CAS-Nummer: 611-17-6
EINECS: 210-257-2
UN: 3265
ADR: 8 II

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR