2-Desoxy-D(-)-ribose ≥99%

Lieferant: Thermo Fisher Scientific

Synonyms: 2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal

A11990.03 A11990.06 A11990.14
A11990.03EA 44.6 CHF
A11990.03 A11990.06 A11990.14
2-Desoxy-D(-)-ribose ≥99%
2-Desoxy-D(-)-ribose

Purity: 99% CAS No.: 533-67-5 Molecular Formula: C5H10O4 Formula Weight: 134.13 Melting Point: 76-85° Beilstein Reference: 1721978 Merck Reference: 14,2908 Optical Rotation: -56° (c=1 in water, 24h) MDL No.: MFCD00135904

Formel: C₅H₁₀O₄
MW: 134.13 g/mol
Siedepunkt: 292
Melting Pt: 76…85 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00135904
CAS-Nummer: 533-67-5
EINECS: 208-573-0
Merck Index: 13,02925

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR