1-Ioddodecan ≥98% stabilisiert

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 1-Dodecyl iodide

A11909.14 A11909.22 A11909.36
A11909.14EA 84.9 CHF
A11909.14 A11909.22 A11909.36
1-Ioddodecan ≥98% stabilisiert
1-Ioddodecan

Stabilized with copper


Purity: 98% CAS No.: 4292-19-7 Molecular Formula: CH3(CH2)11I Formula Weight: 296.24 Flash Point: 110°(230°F) Refractive Index: 1.4844 Melting Point: -3° Boiling Point: 128-130°/11mm Beilstein Reference: 1738943 MDL No.: MFCD00001088

Formel: C₁₂H₂₅I
MW: 296.24 g/mol
Siedepunkt: 128…130 °C (11 mmHg)
Melting Pt: –3 °C
Dichte: 1.201
Flash-Pt: >110 °C (230 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00001088
CAS-Nummer: 4292-19-7
EINECS: 224-293-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR