Zinn(II)oxalat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Tin (II) ethanedioate, Oxalic acid tin (II) salt

A11689.0E A11689.22 A11689.36
A11689.0EEA 734 CHF
A11689.0E A11689.22 A11689.36
Zinn(II)oxalat ≥98%
Zinn(II)oxalat

Purity: 98% CAS No.: 814-94-8 Molecular Formula: C2O4Sn Formula Weight: 206.72 Melting Point: 280° dec. Beilstein Reference: 3708588 Merck Reference: 14,8786 UN No.: 3288 MDL No.: MFCD00040678

Formel: C₂O₄Sn
MW: 206.73 g/mol
Melting Pt: 280 °C (Zers.)
Dichte: 3.56
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00040678
CAS-Nummer: 814-94-8
EINECS: 212-414-0
UN: 3288
ADR: 6.1 III
Merck Index: 13,08865

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR