1-Brom-4-fluorbenzol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 4-Bromofluorobenzene

A11630.0E A11630.22 A11630.36
A11630.0EEA 891 CHF
A11630.0E A11630.22 A11630.36
1-Brom-4-fluorbenzol ≥99%
1-Brom-4-fluorbenzol

Purity: 99% CAS No.: 460-00-4 Molecular Formula: C6H4BrF Formula Weight: 175.01 Flash Point: 60°(140°F) Refractive Index: 1.5270 Odour: Aromatic Melting Point: -16° Boiling Point: 151-153° Beilstein Reference: 774102 UN No.: 1993 MDL No.: MFCD00000342

Formel: C₆H₄BrF
MW: 175 g/mol
Siedepunkt: 151…153 °C
Melting Pt: –16 °C
Dichte: 1.604
Flash-Pt: 60 °C (140 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000342
CAS-Nummer: 460-00-4
EINECS: 207-300-2
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR