1,3-Cyclohexandion ≥97% may cont. up to 1% NaCl

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: Cyclohexane-1,3-dione

A11109.0B A11109.14 A11109.18 A11109.22
A11109.0BEA 889 CHF
A11109.0B A11109.14 A11109.18 A11109.22
1,3-Cyclohexandion ≥97% may cont. up to 1% NaCl
1,3-Cyclohexandion

Purity: 97+% CAS No.: 504-02-9 Molecular Formula: C6H8O2 Formula Weight: 112.13 Storage Temperature: 2°C to 8°C Melting Point: 101-105° Beilstein Reference: 385888 Merck Reference: 14,3178 MDL No.: MFCD00001585

Formel: C₆H₈O₂
MW: 112.13 g/mol
Siedepunkt: 235
Melting Pt: 101…105 °C
Dichte: 1.100
Lager Temperatur: Kühlschrank
MDL Number: MFCD00001585
CAS-Nummer: 504-02-9
EINECS: 207-980-0
Merck Index: 13,03200

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR