2,4,6-Collidin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

A11058.AC A11058.AE A11058.AP
A11058.ACEA 48.5 CHF
A11058.AC A11058.AE A11058.AP
2,4,6-Collidin ≥99%
2,4,6-Collidin

Purity: 99% CAS No.: 108-75-8 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 54°(129°F) Refractive Index: 1.4980 Odour: Unpleasant pH: 9-10 Melting Point: -43° Boiling Point: 171-172° Beilstein Reference: 107283 Merck Reference: 14,9718 UN No.: 1992 MDL No.: MFCD00006338

Formel: C₈H₁₁N
MW: 121.18 g/mol
Siedepunkt: 171…172 °C
Melting Pt: –43 °C
Dichte: 0.916
Flash-Pt: 54 °C (129 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006338
CAS-Nummer: 108-75-8
EINECS: 203-613-3
UN: 1992
ADR: 3 III
Merck Index: 13,09789

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR