Indolin ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 2,3-Dihydroindol

A11000.0B A11000.18 A11000.30 A11000.PG
A11000.0BEA 1100 CHF
A11000.0B A11000.18 A11000.30 A11000.PG
Indolin ≥99%
Indolin

Purity: 99% CAS No.: 496-15-1 Molecular Formula: C8H9N Formula Weight: 119.17 Flash Point: 92°(197°F) Refractive Index: 1.5930 Odour: Odourless Melting Point: -21° Boiling Point: 220-221° Beilstein Reference: 111915 MDL No.: MFCD00005705

Formel: C₈H₉N
Siedepunkt: 220…221 °C
Melting Pt: –21 °C
Dichte: 1.062
Flash-Pt: 92 °C (197 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00005705
CAS-Nummer: 496-15-1
EINECS: 207-816-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR