p-Anisidin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyms: 4-Anisidine, 4-Aminoanisole, 4-Methoxyaniline

A10946.0B A10946.22 A10946.36 AB113749.0500
A10946.0BEA 265 CHF
A10946.0B A10946.22 A10946.36 ABCRAB113749.0500
p-Anisidin ≥99%
p-Anisidin

Purity: 99% CAS No.: 104-94-9 Molecular Formula: C7H9NO Formula Weight: 123.16 Flash Point: 122°(251°F) Melting Point: 56-60° Boiling Point: 240-243° Beilstein Reference: 471556 Merck Reference: 14,667 UN No.: 2811 MDL No.: MFCD00007864

Formel: C₇H₉NO
MW: 123.15 g/mol
Siedepunkt: 240…243 °C
Melting Pt: 56…60 °C
Dichte: 1.089
Flash-Pt: 122 °C (251 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00007864
CAS-Nummer: 104-94-9
EINECS: 203-254-2
UN: 2811
ADR: 6.1 III
Merck Index: 13,00670

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR