3-Fluoranisol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyms: 1-Fluoro-3-methoxybenzene

A10931.09 A10931.14 A10931.18 A10931.30
A10931.09EA 57 CHF
A10931.09 A10931.14 A10931.18 A10931.30
3-Fluoranisol ≥99%
3-Fluoranisol

Purity: 99% CAS No.: 456-49-5 Molecular Formula: C7H7FO Formula Weight: 126.13 Flash Point: 43°(109°F) Refractive Index: 1.4890 Melting Point: -35° Boiling Point: 158-159° Beilstein Reference: 1858895 UN No.: 1993 MDL No.: MFCD00000335

Formel: C₇H₇FO
MW: 126.13 g/mol
Siedepunkt: 158…159 °C
Melting Pt: –35 °C
Dichte: 1.104
Flash-Pt: 43 °C (109 °F)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00000335
CAS-Nummer: 456-49-5
EINECS: 207-267-4
UN: 1993
ADR: 3 III

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR