Azocarmine G, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyms: Rosinduline, Acid red 101, Basalan Carmine GE

M-Clarity™ Qualitätsstufe = MQ100

A1091-10G A1091-25G
A1091-10GEA 223 CHF
A1091-10G A1091-25G
Azocarmine G, Sigma-Aldrich®
Azocarmine G

Standard stain for microscopy. It may be also be used as a reference standard in the determination of azocarmine G in food products and beverages using high performance liquid chromatography coupled with diode array detector (HPLC-DAD).


  • Azocarmine G is a synthetic dye, commonly used in food and beverages to restore their original appearance, in situations where, the color is affected by processing, storage, packaging and distribution
  • Azocarmine G is useful for protein determination in acidic medium. It is indicated by the formation of purple-red complex
  • Azocarmine G may be used as a reference standard for the determination of the analyte in food and beverages using high-performance liquid chromatography coupled with diode-array detector (HPLC-DAD)

Formel: C₂₈H₁₇N₃Na₂O₆S₂
MW: 601.57 g/mol
Lager Temperatur: Room Temperature
MDL Number: MFCD00042002
CAS-Nummer: 25641-18-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d5bdcf491bedf51a7488bb40fdbd9ecf3f0ce54&vid=759bb0885d13f15213dbb3018c50e7c4d73818d6&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance (Form) Powder
Solubility (Colour) Red or Purple
Solubility (Method) 1 mg/ml in water
Uv-Vis Spectrum E1%=210-350 At 510-524Nm In Water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR