Sulphonazo III sodium salt ≥96%

Lieferant: Thermo Fisher Scientific

Synonyms: 2,7-Bis(2-sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonic Acid Tetrasodium Salt

A10486.06 A10486.14
A10486.06EA 164 CHF
A10486.06 A10486.14
Sulphonazo III sodium salt ≥96%
Sulphonazo III sodium salt

CAS No.: 68504-35-8 Molecular Formula: C22H12N4Na4O14S4 Formula Weight: 776.57 Melting Point: ≈300° Beilstein Reference: 1900148 MDL No.: MFCD00003942

Formel: C₂₂H₁₆N₄O₁₄S₄
MW: 776.55 g/mol
Melting Pt: ∼300 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003942
CAS-Nummer: 68504-35-8
EINECS: 270-924-9

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR