L(+)-Histidin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyms: L-3-Imidazol-4-ylalanine, (S)-(+)-Histidine, (S)-2-Amino-3-(4-imidazolyl)propionic acid, H-His-OH, NSC 137773, L-Histidine

A10413.14 A10413.22 A10413.36
A10413.14EA 50 CHF
A10413.14 A10413.22 A10413.36
L(+)-Histidin ≥98%
L(+)-Histidin
Formel: C₆H₉N₃O₂
MW: 155.16 g/mol
Melting Pt: ∼ 281 °C (Zers.)
Dichte: 1.44 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00064315
CAS-Nummer: 71-00-1
EINECS: 200-745-3
Merck Index: 14,4720
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=badfdb36aac0611e5823a7d05e6cba944f6650fa&vid=9e9f9d881a5b3314c7f342312e77e13493901200&prid=web&clid=SAPDAM

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR