4-Amino-3-hydroxybenzoesäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

A10207.06 A10207.14 A10207.22
A10207.06EA 118 CHF
A10207.06 A10207.14 A10207.22
4-Amino-3-hydroxybenzoesäure 98%
4-Amino-3-hydroxybenzoic acid

Purity: 98% CAS No.: 2374-03-0 Molecular Formula: C7H7NO3 Formula Weight: 153.14 Melting Point: ca 215° dec. Beilstein Reference: 509859

Formel: C₇H₇NO₃
MW: 153.14 g/mol
Melting Pt: ∼215 °C (Zers.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00017094
CAS-Nummer: 2374-03-0

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR