Ammoniumacetat

Lieferant: G Bioscienes
Achtung

Synonyms: Acetic acid ammonium salt

A043
A043EA 128 CHF
A043
Ammoniumacetat
Ammoniumacetat
Formel: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: 114
Dichte: 1.17 g/cm³ (20 °C)
Flash-Pt: 136
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00013066
CAS-Nummer: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR