Ammonium ionophore I - cocktail A, Selectophore™ Material für Ionensensoren, Supelco®

Lieferant: Merck

99978-0.1ML-F
99978-0.1ML-FEA 476 CHF
99978-0.1ML-F
Ammonium ionophore I - cocktail A, Selectophore™ Material für Ionensensoren, Supelco®
Ammonium ionophore I - cocktail A

Composition:
Ammonium ionophore I, 6.9 wt. % (09877)
2-Nitrophenyl octyl ether, 92.4 wt. % (73732)
Potassium tetrakis(4-chlorophenyl)borate, 0.7 wt. % (60591)


Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Order Now

Spezifikation Testergebnisse

APPEARANCE (COLOR) Brown to Very Dark Brown
APPEARANCE (FORM) Liquid or Viscous Liquid
REFRACTIVE INDEX N20/D 1.510 - 1.514
INFRARED SPECTRUM CONFORMS TO STRUCTURE
RECOMMENDED RETEST PERIOD 42 MONTHS


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR