Ammoniaklösung 27,0 - 30,0% NH3 basis NF, FCC, strong, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

9726-02EA 122 CHF
9726-02 9726-05 9726-R
Ammoniaklösung 27,0 - 30,0% NH3 basis NF, FCC, strong, J.T.Baker®
Ammoniaklösung

pH adjusters are used to adjust the pH of formulations. This category includes acids that are notoriously difficult to source as truly cGMP-produced and amino acids that most companies do not fully test for EP, BP and JP. Avantor provides all that and more.

Formel: NH₃
MW: 35.05 g/mol
Siedepunkt: 32…37.7 °C (1013 hPa)
Melting Pt: –60…–57.5 °C
Dichte: 0.88…0.91 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00066650
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Order Now

Spezifikation Testergebnisse

Food GMP Manufactured Product Passes Test
Meets N.F. Requirements = 0.02 %
GMP Manufactured Product Passes Test
Meets F.C.C. Requirements 27.0 - 30.0 %
CAUTION: For Manufacturing, processing or repackaging = 5 ppm
Bulk Pharmaceutical Chemical Passes Test
Bulk Food Chemical Passes Test
NF - Identification Passes Test
NF - Nonvolatile Residue ≤0.02 %
NF - Readily Oxidizable Substances Passes Test
NF - Assay (NH₃)(NF) 27.0 - 30.0 %
FCC - Identification Passes Test
FCC - Nonvolatile Residue ≤0.02 %
FCC - Readily Oxidizable Substances Passes Test
FCC - Assay (NH₃)(NF) 27.0 - 30.0 %
FCC - Lead (Pb) (mg/kg) ≤0.5
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR