Carbonate ionophore VII, Selectophore™, Funktion getestet Ionophor, Supelco®

Lieferant: Merck

Synonyms: N,N-Dioctyl-3alpha,12alpha-bis(4-trifluoroacetylbenzoyloxy)-5beta-cholan-24-amide

93206-50MG
93206-50MGEA 441 CHF
93206-50MG
Carbonate ionophore VII, Selectophore™, Funktion getestet Ionophor, Supelco®
Carbonate ionophore VII
Formel: C₅₈H₇₉F₆NO₇
MW: 1016.24 g/mol
CAS-Nummer: 222310-82-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9de381e179cc0bb250db857e3521cd0a091d0339&vid=6ffda7927e8ea5357721001a326f0c80ceffbf66&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR