Natriumazid, BAKER, Laboratory Reagent, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyms: Sodium triazide

9099.0100EA 86.2 CHF
9099.0100
Natriumazid, BAKER, Laboratory Reagent, J.T.Baker®
Natriumazid
Formel: NaN₃
MW: 65.01 g/mol
Melting Pt: 275
Dichte: 1.85 g/cm³ (20 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00003536
CAS-Nummer: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1 II
Merck Index: 13,08653

Order Now

Spezifikation Testergebnisse

Assay min. 99 %
Alkalinity max. 0.1 %
Heavy Metals (as Pb) max. 0.001 %
Insoluble Matter max. 0.05 %
Loss on Drying max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR